Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268779.3 | NP_001255708.1 | 2 - 3 | 350 | 77 | |
GenBank mRNA | AF210702.1 | - | 2 - 3 | 294 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |