Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268779.3 | NP_001255708.1 | 6 - 7 | 1282 | 74 | |
NM_001268780.3 | NP_001255709.1 | 3 - 4 | 563 | 74 | ||
GenBank mRNA | AF210702.1 | - | 6 - 7 | 1226 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |