Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mex-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070165.6 | NP_502566.1 | 4 - 5 | 1224 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |