Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129340.5 | NP_001122812.1 | 12 - 13 | 2496 | 70 | |
NM_001276877.3 | NP_001263806.1 | 8 - 9 | 1918 | 70 | ||
NM_001276878.3 | NP_001263807.1 | 8 - 9 | 1918 | 70 | ||
NM_001276879.3 | NP_001263808.1 | 8 - 9 | 1918 | 70 | ||
NM_001307025.4 | NP_001293954.1 | 12 - 13 | 2496 | 70 | ||
NM_070197.6 | NP_502598.2 | 12 - 13 | 2496 | 70 | ||
NM_070198.5 | NP_502599.2 | 12 - 13 | 2490 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |