Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129340.5 | NP_001122812.1 | 2 - 3 | 307 | 125 | |
NM_001307025.4 | NP_001293954.1 | 2 - 3 | 307 | 125 | ||
NM_070197.6 | NP_502598.2 | 2 - 3 | 307 | 125 | ||
NM_070198.5 | NP_502599.2 | 2 - 3 | 301 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |