Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qui-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307155.3 | NP_001294084.1 | 6 - 7 | 1550 | 67 | |
NM_070212.4 | NP_502613.2 | 11 - 12 | 2357 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |