Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sru-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070216.6 | NP_502617.2 | 1 - 2 | 247 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |