Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276881.3 | NP_001263810.1 | 2 - 3 | 680 | 144 | |
NM_070230.5 | NP_502631.1 | 3 - 4 | 832 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |