Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268838.3 | NP_001255767.1 | 1 - 2 | 276 | 62 | |
NM_001307065.3 | NP_001293994.1 | 1 - 2 | 273 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |