Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268838.3 | NP_001255767.1 | 6 - 7 | 1841 | 73 | |
NM_001268839.3 | NP_001255768.1 | 1 - 2 | 458 | 73 | ||
NM_001307065.3 | NP_001293994.1 | 6 - 7 | 1829 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |