Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkcc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070303.2 | NP_502704.1 | 10 - 11 | 2856 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |