Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070309.3 | NP_502710.2 | 9 - 10 | 2119 | 112 | |
NR_131638.1 | 9 - 10 | 1787 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |