Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqrd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268869.4 | NP_001255798.1 | 6 - 7 | 1189 | 92 | |
NM_001268870.3 | NP_001255799.1 | 6 - 7 | 1053 | 92 | ||
NM_001268871.3 | NP_001255800.1 | 5 - 6 | 697 | 92 | ||
NM_001307012.3 | NP_001293941.1 | 5 - 6 | 844 | 92 | ||
NM_001307302.3 | NP_001294231.1 | 3 - 4 | 298 | 92 | ||
NM_001307303.3 | NP_001294232.1 | 3 - 4 | 445 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |