Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eps-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028350.6 | NP_001023521.2 | 2 - 3 | 414 | 55 | |
NM_001047582.5 | NP_001041047.1 | 4 - 5 | 706 | 55 | ||
NM_001047583.7 | NP_001041048.1 | 4 - 5 | 712 | 55 | ||
NM_001306998.2 | NP_001293927.1 | 4 - 5 | 705 | 55 | ||
NM_001306999.3 | NP_001293928.1 | 3 - 4 | 465 | 55 | ||
NM_001307000.3 | NP_001293929.1 | 2 - 3 | 414 | 55 | ||
NM_001372963.2 | NP_001360610.1 | 4 | 633 | 55 | ||
NM_001372964.2 | NP_001360611.1 | 4 | 633 | 55 | ||
NM_070402.6 | NP_502803.3 | 3 - 4 | 465 | 55 | ||
GenBank mRNA | AY611484.1 | - | 4 - 5 | 630 | 55 | |
AY611485.1 | - | 4 - 5 | 630 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |