Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276894.4 | NP_001263823.2 | 2 - 3 | 184 | 88 | |
NM_001383253.1 | 2 - 3 | 181 | 88 | |||
NR_101983.1 | 2 - 3 | 128 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |