Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268936.4 | NP_001255865.1 | 10 - 11 | 1600 | 74 | |
NM_001268937.3 | NP_001255866.1 | 10 - 11 | 1601 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |