Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070492.7 | NP_502893.2 | 4 - 5 | 848 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |