Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047546.3 | NP_001041011.1 | 2 - 3 | 349 | 132 | |
NM_001307189.3 | NP_001294118.1 | 2 - 3 | 349 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |