Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hlh-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028022.3 | NP_001023193.2 | 1 - 2 | 295 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |