Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rom-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047548.5 | NP_001041013.2 | 1 - 2 | 44 | 144 | |
NM_001083250.6 | NP_001076719.2 | 1 - 2 | 44 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |