Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070729.7 | NP_503130.2 | 5 - 6 | 687 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |