Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313285.3 | NP_001300214.1 | 3 - 4 | 1815 | 96 | |
NM_001313286.3 | NP_001300215.1 | 1 - 2 | 384 | 96 | ||
NM_070750.7 | NP_503151.1 | 5 - 6 | 2107 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |