Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bicd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306805.2 | NP_001293734.1 | 7 - 8 | 1542 | 81 | |
NM_001306806.4 | NP_001293735.1 | 7 - 8 | 1544 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |