Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lec-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028020.5 | NP_001023191.1 | 3 - 4 | 484 | 123 | |
GenBank mRNA | AB038502.2 | - | 3 - 4 | 409 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |