Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380283.2 | NP_001368373.1 | 1 - 2 | 88 | 124 | |
NM_068617.8 | NP_501018.1 | 6 - 7 | 867 | 124 | ||
GenBank mRNA | AF480439.1 | - | 6 - 7 | 862 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |