Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyci-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372852.3 | NP_001360494.1 | 9 | 1536 | 125 | |
NM_001383110.1 | 8 - 9 | 1449 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Both primers and probe map within a single exon |