Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047601.5 | NP_001041066.1 | 3 - 4 | 387 | 68 | |
NM_001380299.1 | NP_001367362.1 | 3 - 4 | 318 | 68 | ||
GenBank mRNA | AF077868.1 | - | 2 - 3 | 301 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |