Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abt-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070774.4 | NP_503175.1 | 8 - 9 | 3106 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |