Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070812.1 | NP_503213.1 | 2 - 3 | 578 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |