Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sru-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313161.1 | NP_001300090.1 | 2 - 3 | 442 | 131 | |
NM_070886.2 | NP_503287.2 | 2 - 3 | 442 | 131 | ||
NR_132481.1 | 2 - 3 | 442 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |