Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313163.4 | NP_001300092.1 | 1 - 2 | 180 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |