Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: icl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026196.7 | NP_001021367.1 | 2 - 3 | 540 | 80 | |
NM_070905.6 | NP_503306.1 | 2 - 3 | 650 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |