Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313217.1 | NP_001300146.1 | 2 - 3 | 408 | 105 | |
NM_070940.2 | NP_503341.2 | 3 - 4 | 516 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |