Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptr-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313272.3 | NP_001300201.1 | 4 | 2241 | 85 | |
NM_001313273.5 | NP_001300202.1 | 7 | 2520 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Amplicon spans exons and probe does not span exons |