Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313185.3 | NP_001300114.1 | 5 - 6 | 1997 | 106 | |
NM_001313186.4 | NP_001300115.1 | 5 | 1663 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |