Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071043.5 | NP_503444.4 | 1 - 2 | 124 | 105 | |
NR_132502.1 | 1 - 2 | 122 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |