Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gst-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269073.2 | NP_001256002.1 | 2 - 3 | 388 | 90 | |
NM_001269074.3 | NP_001256003.1 | 1 - 2 | 163 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |