Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srab-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313374.1 | NP_001300303.1 | 2 - 3 | 202 | 151 | |
NM_071093.2 | NP_503494.1 | 3 - 4 | 468 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |