Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313256.3 | NP_001300185.1 | 1 - 2 | 134 | 68 | |
NM_001313257.3 | NP_001300186.1 | 2 - 3 | 314 | 68 | ||
NM_071134.5 | NP_503535.2 | 4 - 5 | 412 | 68 | ||
GenBank mRNA | EF471978.1 | - | 3 - 4 | 308 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |