Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnt-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276903.3 | NP_001263832.1 | 5 - 6 | 1030 | 91 | |
NM_071143.6 | NP_503544.2 | 5 - 6 | 951 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |