Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313133.3 | NP_001300062.1 | 1 - 2 | 214 | 99 | |
NM_071179.4 | NP_503580.2 | 2 - 3 | 322 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |