Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-33C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316789.1 | NP_001303718.1 | 1 - 2 | 195 | 111 | |
NM_071193.3 | NP_503594.1 | 7 - 8 | 2693 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |