Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071210.5 | NP_503611.3 | 5 - 6 | 852 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |