Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071325.5 | NP_503726.1 | 2 - 3 | 458 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |