Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kel-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313280.3 | NP_001300209.1 | 3 | 223 | 86 | |
NM_071328.8 | NP_503729.4 | 3 - 4 | 255 | 86 | ||
GenBank mRNA | DQ338465.1 | - | 2 - 3 | 112 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |