Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ketn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307737.5 | NP_001294666.1 | 16 - 17 | 5070 | 65 | |
NM_001307738.4 | NP_001294667.1 | 16 - 17 | 5070 | 65 | ||
NM_001307739.4 | NP_001294668.1 | 16 - 17 | 5070 | 65 | ||
NM_001307740.4 | NP_001294669.1 | 16 - 17 | 5070 | 65 | ||
NM_001316807.3 | NP_001303736.1 | 3 - 4 | 594 | 65 | ||
NM_001377590.2 | NP_001364781.1 | 8 - 9 | 3153 | 65 | ||
NM_001377591.2 | NP_001364828.1 | 8 - 9 | 3153 | 65 | ||
NM_001377592.1 | NP_001364782.1 | 8 - 9 | 3153 | 65 | ||
NM_001377593.1 | NP_001364783.1 | 8 - 9 | 3153 | 65 | ||
NM_001377594.2 | NP_001364827.1 | 8 - 9 | 3153 | 65 | ||
GenBank mRNA | AY819766.1 | - | 11 - 12 | 3940 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |