Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071383.1 | NP_503784.1 | 4 - 5 | 864 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |