Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316787.1 | NP_001303716.1 | 2 - 3 | 264 | 122 | |
NM_001316788.1 | NP_001303717.1 | 3 - 4 | 453 | 122 | ||
NM_071392.1 | NP_503793.1 | 4 - 5 | 756 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |