Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071427.6 | NP_503828.3 | 2 - 3 | 774 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |