Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071433.2 | NP_503834.2 | 2 - 3 | 621 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |